missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 3-Methyladenine, 90+%

Product Code. 10096372 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass Bottle
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 10096372

Brand: Thermo Scientific Chemicals 379791000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5142-23-4 | C6H7N5 | 149.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5142-23-4
Molecular Formula C6H7N5
Molecular Weight (g/mol) 149.16
MDL Number MFCD00010531
InChI Key FSASIHFSFGAIJM-UHFFFAOYSA-N
Synonym 3-methyladenine, 3-methyl-3h-purin-6-amine, 6-amino-3-methylpurine, 3-ma, 3h-purin-6-amine, 3-methyl, 3-methyl-3h-adenine, n3-methyladenine, adenine, 3-methyl, 3-methyl-3h-purin-6-ylamine, 3-methyladenine 3-ma
PubChem CID 1673
ChEBI CHEBI:38635
SMILES CN1C=NC(N)=C2N=CN=C12

Specifications

CAS Min % 90.0
CAS Max % 100.0
Melting Point 300°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 90% min. (HPLC)
Packaging Glass Bottle
Quantity 100 mg
Beilstein 26,V,17,151
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,ethanol,dmso,chloroform
IUPAC Name 3-methyl-3H-purin-6-amine
Formula Weight 149.15
Percent Purity 90+%
Physical Form Powder
Chemical Name or Material 3-Methyladenine
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methyladenine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.