Learn More
(3-Methylaminopropyl)trimethoxysilane, 95%, Thermo Scientific Chemicals
£83.42 - £251.25
Chemical Identifiers
CAS | 3069-25-8 |
---|---|
Molecular Formula | C7H19NO3Si |
Molecular Weight (g/mol) | 193.32 |
MDL Number | MFCD00048021 |
InChI Key | DVYVMJLSUSGYMH-UHFFFAOYSA-N |
Synonym | n-methylaminopropyltrimethoxysilane, n-methyl-3-trimethoxysilyl propan-1-amine, trimethoxy 3-methylamino propyl silane, 1-propanamine, n-methyl-3-trimethoxysilyl, n-methyl-3-trimethoxysilyl propylamine, n-methyl-3-aminopropyltrimethoxysilane, 3-methylaminopropyl-trimethoxysilane, methyl 3-trimethoxysilyl propyl amine, n-methyl-3-aminopropyl-trimethoxysilane, acmc-20ak27 |
PubChem CID | 76480 |
IUPAC Name | N-methyl-3-trimethoxysilylpropan-1-amine |
SMILES | CNCCC[Si](OC)(OC)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11379225
|
Thermo Scientific Alfa Aesar
L16541.14 |
25 g |
£83.42
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11389225
|
Thermo Scientific Alfa Aesar
L16541.22 |
100 g |
£251.25
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
(3-Methylaminopropyl)trimethoxysilane is used as coupling agent, adhesion promoters
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(3-Methylaminopropyl)trimethoxysilane is used as coupling agent, adhesion promoters
Solubility
Hydrolyzes in water.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
3069-25-8 | |
193.32 | |
DVYVMJLSUSGYMH-UHFFFAOYSA-N | |
76480 | |
CNCCC[Si](OC)(OC)OC |
C7H19NO3Si | |
MFCD00048021 | |
n-methylaminopropyltrimethoxysilane, n-methyl-3-trimethoxysilyl propan-1-amine, trimethoxy 3-methylamino propyl silane, 1-propanamine, n-methyl-3-trimethoxysilyl, n-methyl-3-trimethoxysilyl propylamine, n-methyl-3-aminopropyltrimethoxysilane, 3-methylaminopropyl-trimethoxysilane, methyl 3-trimethoxysilyl propyl amine, n-methyl-3-aminopropyl-trimethoxysilane, acmc-20ak27 | |
N-methyl-3-trimethoxysilylpropan-1-amine |
Specifications
106°C (30mmHg) | |
C7H19NO3Si | |
2072720 | |
n-methylaminopropyltrimethoxysilane, n-methyl-3-trimethoxysilyl propan-1-amine, trimethoxy 3-methylamino propyl silane, 1-propanamine, n-methyl-3-trimethoxysilyl, n-methyl-3-trimethoxysilyl propylamine, n-methyl-3-aminopropyltrimethoxysilane, 3-methylaminopropyl-trimethoxysilane, methyl 3-trimethoxysilyl propyl amine, n-methyl-3-aminopropyl-trimethoxysilane, acmc-20ak27 | |
DVYVMJLSUSGYMH-UHFFFAOYSA-N | |
N-methyl-3-trimethoxysilylpropan-1-amine | |
76480 | |
95% | |
Moisture sensitive | |
(3-Methylaminopropyl)trimethoxysilane |
3069-25-8 | |
MFCD00048021 | |
82°C (179°F) | |
Hydrolyzes in water. | |
CNCCC[Si](OC)(OC)OC | |
193.32 | |
193.32 | |
1.419 | |
0.978 |
Safety and Handling
GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
P210-P280-P305+P351+P338-P310-P370+P378q-P501c
H227-H318
missing translation for 'einecsNumber' : 221-334-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- (3-Methylaminopropyl)trimethoxysilane
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : (3-Methylaminopropyl)trimethoxysilane
RUO – Research Use Only