missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzeneboronic acid, 97%, Thermo Scientific Chemicals

£40.30 - £371.00

Chemical Identifiers

CAS 17933-03-8
Molecular Formula C7H9BO2
Molecular Weight (g/mol) 135.96
MDL Number MFCD00040198
InChI Key BJQCPCFFYBKRLM-UHFFFAOYSA-N
Synonym 3-tolylboronic acid, m-tolylboronic acid, 3-methylphenyl boronic acid, 3-methylbenzeneboronic acid, 3-methyl phenyl boronic acid, 3-tolyboronic acid, 3-methylphenyl boranediol, m-methylphenylboronic acid, m-tolyl boronic acid
PubChem CID 2733950
IUPAC Name (3-methylphenyl)boronic acid
SMILES CC1=CC=CC(=C1)B(O)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11456206
View Documents
Thermo Scientific Alfa Aesar
B23025.03
1 g
£40.30
1g
Please to purchase this item. Need a web account? Register with us today!
11466206
View Documents
Thermo Scientific Alfa Aesar
B23025.06
5 g
£83.20
5g
Please to purchase this item. Need a web account? Register with us today!
11476206
View Documents
Thermo Scientific Alfa Aesar
B23025.14
25 g
£371.00
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Suzuki coupling reactions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

17933-03-8
135.96
BJQCPCFFYBKRLM-UHFFFAOYSA-N
2733950
CC1=CC=CC(=C1)B(O)O
C7H9BO2
MFCD00040198
3-tolylboronic acid, m-tolylboronic acid, 3-methylphenyl boronic acid, 3-methylbenzeneboronic acid, 3-methyl phenyl boronic acid, 3-tolyboronic acid, 3-methylphenyl boranediol, m-methylphenylboronic acid, m-tolyl boronic acid
(3-methylphenyl)boronic acid

Specifications

17933-03-8
C7H9BO2
2935968
BJQCPCFFYBKRLM-UHFFFAOYSA-N
(3-methylphenyl)boronic acid
2733950
97%
∼160°C to 162°C
MFCD00040198
3-tolylboronic acid, m-tolylboronic acid, 3-methylphenyl boronic acid, 3-methylbenzeneboronic acid, 3-methyl phenyl boronic acid, 3-tolyboronic acid, 3-methylphenyl boranediol, m-methylphenylboronic acid, m-tolyl boronic acid
CC1=CC=CC(=C1)B(O)O
135.96
135.96
3-Methylbenzeneboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 000-000-0

missing translation for 'rtecsNumber' : ED7788888

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only