missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzenecarboximidamide hydrochloride, 95%, Thermo Scientific™

Product Code. 10783833
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 20680-59-5
Molecular Formula C8H11ClN2
Molecular Weight (g/mol) 170.64
MDL Number MFCD02180878
InChI Key QEAXZIMXYPAZAX-UHFFFAOYSA-N
Synonym 3-methylbenzenecarboximidamide hydrochloride, 3-methylbenzimidamide hydrochloride, 3-methyl-benzamidine hydrochloride, 3-methylbenzene-1-carboximidamide hydrochloride, benzenecarboximidamide, 3-methyl-, monohydrochloride, m-toluamidine hydrochloride, 3-methylbenzamidine, hydrochloride, 3-methylbenzenecarboxamidine, chloride, 3-methylbenzenecarboximidamide hcl, 3-carbamimidoyltoluene hydrochloride
PubChem CID 2800701
IUPAC Name 3-methylbenzenecarboximidamide;hydrochloride
SMILES CC1=CC=CC(=C1)C(=N)N.Cl

Specifications

CAS Min % 97.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Linear Formula CH3C6H4C(=NH)NH2·HCl
Quantity 1 g
Formula Weight 170.64
Percent Purity 97%
Chemical Name or Material 3-Methylbenzenecarboximidamide hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.