missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylcarbamoyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals

£58.20 - £198.00

Chemical Identifiers

CAS 832695-88-2
Molecular Formula C8H10BNO3
Molecular Weight (g/mol) 178.98
MDL Number MFCD04038918
InChI Key FYFFPNFUVMBPRZ-UHFFFAOYSA-N
Synonym 3-n-methylaminocarbonyl phenylboronic acid, 3-methylcarbamoyl phenyl boronic acid, 3-methylcarbamoyl benzeneboronic acid, 3-methylcarbamoyl phenylboronic acid, 3-n-methylaminocarbonyl benzeneboronic acid, 3-borono-n-methylbenzamide, boronic acid, 3-methylamino carbonyl phenyl, acmc-209prq, ksc496e8l
PubChem CID 2773524
IUPAC Name [3-(methylcarbamoyl)phenyl]boronic acid
SMILES CNC(=O)C1=CC=CC(=C1)B(O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15429907
View Documents
Thermo Scientific Alfa Aesar
H53141.MD
250 mg
£58.20
250mg
Please to purchase this item. Need a web account? Register with us today!
15419907
View Documents
Thermo Scientific Alfa Aesar
H53141.03
1 g
£198.00
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

832695-88-2
178.98
FYFFPNFUVMBPRZ-UHFFFAOYSA-N
2773524
CNC(=O)C1=CC=CC(=C1)B(O)O
C8H10BNO3
MFCD04038918
3-n-methylaminocarbonyl phenylboronic acid, 3-methylcarbamoyl phenyl boronic acid, 3-methylcarbamoyl benzeneboronic acid, 3-methylcarbamoyl phenylboronic acid, 3-n-methylaminocarbonyl benzeneboronic acid, 3-borono-n-methylbenzamide, boronic acid, 3-methylamino carbonyl phenyl, acmc-209prq, ksc496e8l
[3-(methylcarbamoyl)phenyl]boronic acid

Specifications

832695-88-2
C8H10BNO3
3-n-methylaminocarbonyl phenylboronic acid, 3-methylcarbamoyl phenyl boronic acid, 3-methylcarbamoyl benzeneboronic acid, 3-methylcarbamoyl phenylboronic acid, 3-n-methylaminocarbonyl benzeneboronic acid, 3-borono-n-methylbenzamide, boronic acid, 3-methylamino carbonyl phenyl, acmc-209prq, ksc496e8l
CNC(=O)C1=CC=CC(=C1)B(O)O
178.98
178.98
3-(Methylcarbamoyl)benzeneboronic acid
114°C to 124°C
MFCD04038918
FYFFPNFUVMBPRZ-UHFFFAOYSA-N
[3-(methylcarbamoyl)phenyl]boronic acid
2773524
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only