missing translation for 'onlineSavingsMsg'
Learn More

3-Methylcyclohexanone, 97%, Thermo Scientific Chemicals

Product Code. 10366480
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-24-2
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00001639
InChI Key UJBOOUHRTQVGRU-UHFFFAOYNA-N
Synonym 3-methylcyclohexanone, cyclohexanone, 3-methyl, methyl-3 cyclohexanone-1, 3-methyl cyclohexanone, methyl-3 cyclohexanone-1 french, m-methylcyclohexanone, r-3-methylcyclohexanone, +-3r-3-methylcyclohexanone, cyclohexanone, 3-methyl-, r, cyclohexanone, 3-methyl-, s
PubChem CID 11567
IUPAC Name 3-methylcyclohexan-1-one
SMILES CC1CCCC(=O)C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -73.0°C
Color Colorless to Yellow
Density 0.9150g/mL
Boiling Point 169.0°C to 170.0°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3C6H9(=O)
Refractive Index 1.4440 to 1.4460
Quantity 5 g
Beilstein 07, 15
Specific Gravity 0.915
Solubility Information Solubility in water: insoluble
Formula Weight 112.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Methylcyclohexanone, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Methylcyclohexanone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'