missing translation for 'onlineSavingsMsg'
Learn More

3-Methylcyclopentane-1,2-dione, 98+%, Thermo Scientific Chemicals

Product Code. 11389459
Click to view available options
Quantity:
10 g
50 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 765-70-8
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00001417
InChI Key OACYKCIZDVVNJL-UHFFFAOYSA-N
Synonym 3-methyl-1,2-cyclopentanedione, 1,2-cyclopentanedione, 3-methyl, methylcyclopentenolone diketo form, fema no. 2700, methylcyclopentenolone natural, kentonarome, benzil-related compound, 45, 3-methyl-1,2-cyclopentanedione maple lactone, maple lactone mcp, methyl-cyclopentenolone
PubChem CID 61209
IUPAC Name 3-methylcyclopentane-1,2-dione
SMILES CC1CCC(=O)C1=O

Specifications

Melting Point 103°C to 107°C
Quantity 10 g
Beilstein 2076520
Formula Weight 112.13
Percent Purity ≥98%
Chemical Name or Material 3-Methylcyclopentane-1,2-dione

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'