missing translation for 'onlineSavingsMsg'
Learn More

3-Methylindole, 98%, Thermo Scientific Chemicals

Product Code. 10553481
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-34-1
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.18
MDL Number MFCD00005627
InChI Key ZFRKQXVRDFCRJG-UHFFFAOYSA-N
Synonym 3-methylindole, skatole, scatole, skatol, 1h-indole, 3-methyl, beta-methylindole, indole, 3-methyl, 3-mi, 3-methyl-4,5-benzopyrrole, 3-methyl indole
PubChem CID 6736
ChEBI CHEBI:9171
IUPAC Name 3-methyl-1H-indole
SMILES CC1=CNC2=CC=CC=C12

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 95.0°C to 98.0°C
Color Brown to White
Boiling Point 265.0°C to 266.0°C (755.0 mmHg)
Flash Point 132°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Quantity 25 g
Beilstein 20, 315
Merck Index 15, 8698
Solubility Information Solubility in water: 0.45g/L (20°C). Other solubilities: soluble in hot water,soluble in alcohol,benzene,chloroform and ether
Formula Weight 131.18
Percent Purity 98%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material 3-Methylindole, 98%
Show More Show Less
Product Identifier
  • 3-Methylindole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'