missing translation for 'onlineSavingsMsg'
Learn More

3-Methylisoxazole-5-carbonyl chloride, 95%, Thermo Scientific™

Product Code. 10258233
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10258233

Brand: Thermo Scientific Maybridge CC27002CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 49783-72-4
Molecular Formula C5H4ClNO2
Molecular Weight (g/mol) 145.54
MDL Number MFCD09817464
InChI Key VUBCNGCMOPTDED-UHFFFAOYSA-N
Synonym 3-methylisoxazole-5-carbonyl chloride, 3-methyl-5-isoxazolecarbonyl chloride, 5-isoxazolecarbonyl chloride, 3-methyl, aronis23810, 3-methylisoxazol-5-carbonyl chloride, 3-methyl-isoxazole-5-carbonyl chloride, 5-isoxazolecarbonyl chloride, 3-methyl-9ci
PubChem CID 12905099
IUPAC Name 3-methyl-1,2-oxazole-5-carbonyl chloride
SMILES CC1=NOC(=C1)C(Cl)=O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 145.54
Percent Purity 95%
Chemical Name or Material 3-Methylisoxazole-5-carbonyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.