missing translation for 'onlineSavingsMsg'
Learn More

3-Methylphenylzinc iodide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Product Code. 15415365
Click to view available options
Quantity:
50 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 312693-24-6
Density 0.984
Molecular Formula C7H7IZn
MDL Number MFCD01311453
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive
Synonym zinc,iodo 3-methylphenyl, 3-methylphenyl zincylium iodide
InChI Key GFDZNGFKUKUHTR-UHFFFAOYSA-M
SMILES CC1=C[C-]=CC=C1.[Zn+]I
IUPAC Name iodozinc(1+);methylbenzene
Molecular Weight (g/mol) 283.417
PubChem CID 3614645
Formula Weight 283.42
Chemical Name or Material 3-Methylphenylzinc iodide, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'