missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylsulfonyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals

Product Code. 15479687
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 373384-18-0
Molecular Formula C7H9BO4S
Molecular Weight (g/mol) 200.015
MDL Number MFCD03092935
InChI Key HZFFUMBZBGETES-UHFFFAOYSA-N
Synonym 3-methylsulfonyl phenylboronic acid, 3-methylsulfonyl phenyl boronic acid, 3-methylsulfonylphenyl boronic acid, 3-methanesulfonyl phenylboronic acid, 3-methanesulfonyl phenyl boronic acid, 3-methanesulfonylphenylboronic acid, boronic acid, 3-methylsulfonyl phenyl, 3-methanesulfonylphenyl boronic acid, 3-methanesulphonyl phenylboronic acid
PubChem CID 2773542
IUPAC Name (3-methylsulfonylphenyl)boronic acid
SMILES B(C1=CC(=CC=C1)S(=O)(=O)C)(O)O

Specifications

Melting Point 102°C to 110°C
Quantity 5 g
Formula Weight 200.02
Percent Purity 98%
Chemical Name or Material 3-(Methylsulfonyl)benzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'