missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Methylsulfonylamino)benzeneboronic acid, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Benzeneboronic acid mediates the ortho specific alpha hydroxyalkylation of phenols by aldehydes. The chemical itself can be used as a template for Diels-Alder reaction by forming linkage with a hydroxy diene and a hydroxy dieneophile.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Benzeneboronic acid mediates the ortho specific alpha hydroxyalkylation of phenols by aldehydes. The chemical itself can be used as a template for Diels-Alder reaction by forming linkage with a hydroxy diene and a hydroxy dieneophile.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Keep Cold
Benzeneboronic acid mediates the ortho specific alpha hydroxyalkylation of phenols by aldehydes. The chemical itself can be used as a template for Diels-Alder reaction by forming linkage with a hydroxy diene and a hydroxy dieneophile.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Keep Cold
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 90°C to 96°C |
Quantity | 5 g |
Formula Weight | 215.03 |
Percent Purity | 97% |
Chemical Name or Material | 3-(Methylsulfonylamino)benzeneboronic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'