missing translation for 'onlineSavingsMsg'
Learn More

3-Methylthio-1-propanol, 98%, Thermo Scientific Chemicals

Product Code. 11418593
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 505-10-2
Molecular Formula C4H10OS
Molecular Weight (g/mol) 106.18
MDL Number MFCD00036560
InChI Key CZUGFKJYCPYHHV-UHFFFAOYSA-N
Synonym methionol, 3-methylthiopropanol, 3-methylthio-1-propanol, 3-hydroxypropyl methyl sulfide, 3-methylthio propanol, 3-methylmercapto-1-propanol, 1-propanol, 3-methylthio, 3-methylsulfanyl propan-1-ol, gamma-methylmercaptopropyl alcohol
PubChem CID 10448
ChEBI CHEBI:49019
IUPAC Name 3-methylsulfanylpropan-1-ol
SMILES CSCCCO

Specifications

Density 1.027
Boiling Point 89°C to 90°C (13 mmHg)
Flash Point 90°C (195°F)
Refractive Index 1.4904
Quantity 5 g
Formula Weight 106.19
Percent Purity 98%
Chemical Name or Material 3-Methylthio-1-propanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'