missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Methylthio)phenyl isocyanate, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
3-(Methylthio)phenyl isocyanate is used as a coupling reagent, N-protecting reagent and as gas chromatography derivatizing reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-(Methylthio)phenyl isocyanate is used as a coupling reagent, N-protecting reagent and as gas chromatography derivatizing reagent.
Solubility
Hydrolyzes in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents, amines and alcohols. It is sensitive to moisture. Store under dry inert gas.
3-(Methylthio)phenyl isocyanate is used as a coupling reagent, N-protecting reagent and as gas chromatography derivatizing reagent.
Solubility
Hydrolyzes in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents, amines and alcohols. It is sensitive to moisture. Store under dry inert gas.
Specifications
Specifications
Density | 1.199 |
Boiling Point | 129°C to 130°C (5 mmHg) |
Flash Point | 113°C (235°F) |
Refractive Index | 1.603 |
Quantity | 1 g |
UN Number | UN2206 |
Beilstein | 387507 |
Sensitivity | Moisture sensitive |
Solubility Information | Hydrolyzes in water. |
Formula Weight | 165.22 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'