missing translation for 'onlineSavingsMsg'
Learn More

3-Methylthiophene-2-carbonyl chloride, 98%, Thermo Scientific™

£31.50 - £101.00

Chemical Identifiers

CAS 61341-26-2
Molecular Formula C6H5ClOS
Molecular Weight (g/mol) 160.62
MDL Number MFCD00130091
InChI Key AJSVPEVDFBYRCH-UHFFFAOYSA-N
Synonym 3-methyl-2-thiophenecarbonyl chloride, 3-methylthiophene-2-carbonylchloride, 3-methyl-2-thenoyl chloride, 2-3-methyl thiophene carboxylicacid chloride, pubchem5561, acmc-209mrl, 3-methylthenoyl chloride, 3-methyl-2-thenoylchloride, 3-methyl-2-thienoyl chloride, 3-methylthiophenecarbonylchloride
PubChem CID 2737605
IUPAC Name 3-methylthiophene-2-carbonyl chloride
SMILES CC1=C(SC=C1)C(Cl)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11354865
View Documents
Thermo Scientific Alfa Aesar
L17989.04
2 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11364865
View Documents
Thermo Scientific Alfa Aesar
L17989.09
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

61341-26-2
160.62
AJSVPEVDFBYRCH-UHFFFAOYSA-N
2737605
CC1=C(SC=C1)C(Cl)=O
C6H5ClOS
MFCD00130091
3-methyl-2-thiophenecarbonyl chloride, 3-methylthiophene-2-carbonylchloride, 3-methyl-2-thenoyl chloride, 2-3-methyl thiophene carboxylicacid chloride, pubchem5561, acmc-209mrl, 3-methylthenoyl chloride, 3-methyl-2-thenoylchloride, 3-methyl-2-thienoyl chloride, 3-methylthiophenecarbonylchloride
3-methylthiophene-2-carbonyl chloride

Specifications

61341-26-2
C6H5ClOS
MFCD00130091
116181
3-methyl-2-thiophenecarbonyl chloride, 3-methylthiophene-2-carbonylchloride, 3-methyl-2-thenoyl chloride, 2-3-methyl thiophene carboxylicacid chloride, pubchem5561, acmc-209mrl, 3-methylthenoyl chloride, 3-methyl-2-thenoylchloride, 3-methyl-2-thienoyl chloride, 3-methylthiophenecarbonylchloride
CC1=C(SC=C1)C(Cl)=O
160.62
160.62
3-Methylthiophene-2-carbonyl chloride
216°C to 220°C
1.5855
UN3265
Moisture sensitive
AJSVPEVDFBYRCH-UHFFFAOYSA-N
3-methylthiophene-2-carbonyl chloride
2737605
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 000-000-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications