missing translation for 'onlineSavingsMsg'
Learn More

3-morpholin-4-ylaniline, 97%, Thermo Scientific™

Product Code. 10173714
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 159724-40-0
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.24
MDL Number MFCD03197165
InChI Key ZJWLMZURLIHVHE-UHFFFAOYSA-N
Synonym 3-4-morpholinyl aniline, 3-morpholinoaniline, 3-morpholinobenzenamine, 3-morpholin-4-yl aniline, benzenamine, 3-4-morpholinyl, 3-morpholin-4-yl-phenylamine, 4-3-aminophenyl morpholine, 3-morpholin-4-ylphenylamine, 3-morpholinylaniline, meta-morpholino aniline
PubChem CID 847768
IUPAC Name 3-morpholin-4-ylaniline
SMILES NC1=CC=CC(=C1)N1CCOCC1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 178.23
Percent Purity 97%
Chemical Name or Material 3-Morpholin-4-ylaniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'