missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals

£24.72 - £78.39

Chemical Identifiers

CAS 99-61-6
Molecular Formula C7H5NO3
Molecular Weight (g/mol) 151.12
MDL Number MFCD00007249
InChI Key ZETIVVHRRQLWFW-UHFFFAOYSA-N
Synonym m-nitrobenzaldehyde, benzaldehyde, 3-nitro, 3-formylnitrobenzene, meta-nitrobenzaldehyde, benzaldehyde, m-nitro, 5-nitrobenzaldehyde, unii-g4o92ko71z, 3-nitro-benzaldehyde, ccris 1784, m-nitro-benzaldehyde
PubChem CID 7449
IUPAC Name 3-nitrobenzaldehyde
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10123660
View Documents
Thermo Scientific Acros
128411000
100 g Plastic bottle
£24.72
100g
Please to purchase this item. Need a web account? Register with us today!
10051440
View Documents Promotion Details
Thermo Scientific Acros
128415000
500 g Plastic bottle
£78.39
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

99-61-6
151.12
ZETIVVHRRQLWFW-UHFFFAOYSA-N
7449
C1=CC(=CC(=C1)[N+](=O)[O-])C=O
C7H5NO3
MFCD00007249
m-nitrobenzaldehyde, benzaldehyde, 3-nitro, 3-formylnitrobenzene, meta-nitrobenzaldehyde, benzaldehyde, m-nitro, 5-nitrobenzaldehyde, unii-g4o92ko71z, 3-nitro-benzaldehyde, ccris 1784, m-nitro-benzaldehyde
3-nitrobenzaldehyde

Specifications

99-61-6
100.0
Brown-Yellow to Yellow
Authentic
C7H5NO3
MFCD00007249
14, 6587
Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, chloroformm and ether
C1=CC(=CC(=C1)[N+](=O)[O-])C=O
151.12
151.12
Granular Powder
98.5
56.0°C to 59.0°C
285.0°C to 290.0°C
98.5% min. (GC)
O2NC6H4CHO
07, 250
m-nitrobenzaldehyde, benzaldehyde, 3-nitro, 3-formylnitrobenzene, meta-nitrobenzaldehyde, benzaldehyde, m-nitro, 5-nitrobenzaldehyde, unii-g4o92ko71z, 3-nitro-benzaldehyde, ccris 1784, m-nitro-benzaldehyde
ZETIVVHRRQLWFW-UHFFFAOYSA-N
3-nitrobenzaldehyde
7449
99%
3-Nitrobenzaldehyde, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 202-772-6

missing translation for 'rtecsNumber' : CU7250000

missing translation for 'tsca' : TSCA

Product Identifier
  • 3-Nitrobenzaldehyde

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only