Learn More
3-Nitrophenol, 98+%, Thermo Scientific Chemicals
£23.60 - £381.00
Chemical Identifiers
CAS | 554-84-7 |
---|---|
Molecular Formula | C6H5NO3 |
Molecular Weight (g/mol) | 139.11 |
MDL Number | MFCD00007240 |
InChI Key | RTZZCYNQPHTPPL-UHFFFAOYSA-N |
Synonym | m-nitrophenol, 3-hydroxynitrobenzene, m-hydroxynitrobenzene, phenol, 3-nitro, phenol, m-nitro, m-nitrofenol, meta-nitrophenol, 3-nitro-phenol, m-nitrofenol czech, 1-hydroxy-3-nitrobenzene |
PubChem CID | 11137 |
ChEBI | CHEBI:34346 |
IUPAC Name | 3-nitrophenol |
SMILES | OC1=CC=CC(=C1)[N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11418196
|
Thermo Scientific Alfa Aesar
A15312.09 |
10 g |
£23.60
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11428196
|
Thermo Scientific Alfa Aesar
A15312.18 |
50 g |
£78.90
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11438196
|
Thermo Scientific Alfa Aesar
A15312.30 |
250 g |
£381.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-Nitrophenol is used as indicator in 0.3% Soln in 50% alcohol ph: 6.8 Colorless, 8.6 Yellow. It is used in the synthesis of RAGE inhibitors via the etherification of phenols. Also used in the synthesis of kinase inhibitors against epidermal growth factor receptor T790M mutation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Nitrophenol is used as indicator in 0.3% Soln in 50% alcohol ph: 6.8 Colorless, 8.6 Yellow. It is used in the synthesis of RAGE inhibitors via the etherification of phenols. Also used in the synthesis of kinase inhibitors against epidermal growth factor receptor T790M mutation.
Solubility
Soluble in water (30 mg/ml), ether, alcohol, acetone, and ethanol (700 mg/ml).
Notes
Store in a tightly closed container. Keep from contact with oxidizing materials. Store in a cool, dry, well-ventilated area away from incompatible substances.
Chemical Identifiers
554-84-7 | |
139.11 | |
RTZZCYNQPHTPPL-UHFFFAOYSA-N | |
11137 | |
3-nitrophenol |
C6H5NO3 | |
MFCD00007240 | |
m-nitrophenol, 3-hydroxynitrobenzene, m-hydroxynitrobenzene, phenol, 3-nitro, phenol, m-nitro, m-nitrofenol, meta-nitrophenol, 3-nitro-phenol, m-nitrofenol czech, 1-hydroxy-3-nitrobenzene | |
CHEBI:34346 | |
OC1=CC=CC(=C1)[N+]([O-])=O |
Specifications
554-84-7 | |
1.49 | |
>100°C (212°F) | |
MFCD00007240 | |
1907946 | |
m-nitrophenol, 3-hydroxynitrobenzene, m-hydroxynitrobenzene, phenol, 3-nitro, phenol, m-nitro, m-nitrofenol, meta-nitrophenol, 3-nitro-phenol, m-nitrofenol czech, 1-hydroxy-3-nitrobenzene | |
RTZZCYNQPHTPPL-UHFFFAOYSA-N | |
3-nitrophenol | |
11137 | |
139.11 | |
3-Nitrophenol |
93°C to 98°C | |
194°C (70 mmHg) | |
C6H5NO3 | |
UN1663 | |
14,6619 | |
Soluble in water (30mg/ml),ether,alcohol,acetone,and ethanol (700mg/ml). | |
OC1=CC=CC(=C1)[N+]([O-])=O | |
139.11 | |
CHEBI:34346 | |
≥98% |
Safety and Handling
GHS H Statement
H318-H373-H302-H312-H335-H315
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
P260-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P314-P330-P332+P313-P362-P501c
H302-H315-H318-H373-H500
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROPHENOLS
missing translation for 'einecsNumber' : 209-073-5
missing translation for 'rtecsNumber' : SM1925000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only