missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Nitrophenol, 99%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 554-84-7 |
---|---|
Molecular Formula | C6H5NO3 |
Molecular Weight (g/mol) | 139.11 |
MDL Number | MFCD00007240 |
InChI Key | RTZZCYNQPHTPPL-UHFFFAOYSA-N |
Synonym | m-nitrophenol, 3-hydroxynitrobenzene, m-hydroxynitrobenzene, phenol, 3-nitro, phenol, m-nitro, m-nitrofenol, meta-nitrophenol, 3-nitro-phenol, m-nitrofenol czech, 1-hydroxy-3-nitrobenzene |
PubChem CID | 11137 |
ChEBI | CHEBI:34346 |
IUPAC Name | 3-nitrophenol |
SMILES | OC1=CC=CC(=C1)[N+]([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10387780
|
Thermo Scientific Acros
172300100 |
10 g | Glass bottle |
N/A
|
N/A | ||||
10531431
|
Thermo Scientific Acros
172300500 |
50 g | Glass bottle |
N/A
|
N/A | ||||
11343433
|
Thermo Scientific
172302500 |
250 g | Glass bottle |
N/A
|
N/A | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
554-84-7 | |
139.11 | |
RTZZCYNQPHTPPL-UHFFFAOYSA-N | |
11137 | |
3-nitrophenol |
C6H5NO3 | |
MFCD00007240 | |
m-nitrophenol, 3-hydroxynitrobenzene, m-hydroxynitrobenzene, phenol, 3-nitro, phenol, m-nitro, m-nitrofenol, meta-nitrophenol, 3-nitro-phenol, m-nitrofenol czech, 1-hydroxy-3-nitrobenzene | |
CHEBI:34346 | |
OC1=CC=CC(=C1)[N+]([O-])=O |
Specifications
554-84-7 | |
Yellow to Brown | |
194.0°C (70.0 mmHg) | |
98.5% min. (GC) | |
O2NC6H4OH | |
06,222 | |
1.49 | |
Solubility in water: 13.5g/L (25°C). Other solubilities: soluble 700g/L in ethanol (20°C),soluble in benzene,diethyl ether,alcohol,,hot and diluted solutions,insoluble in petroleum ehter | |
OC1=CC=CC(=C1)[N+]([O-])=O | |
139.11 | |
CHEBI:34346 | |
99% | |
3-Nitrophenol |
94.0°C to 98.0°C | |
1.4900g/mL | |
Authentic | |
C6H5NO3 | |
MFCD00007240 | |
15,6706 | |
m-nitrophenol, 3-hydroxynitrobenzene, m-hydroxynitrobenzene, phenol, 3-nitro, phenol, m-nitro, m-nitrofenol, meta-nitrophenol, 3-nitro-phenol, m-nitrofenol czech, 1-hydroxy-3-nitrobenzene | |
RTZZCYNQPHTPPL-UHFFFAOYSA-N | |
3-nitrophenol | |
11137 | |
139.11 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear prote
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 209-073-5
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 3-Nitrophenol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
RUO – Research Use Only