missing translation for 'onlineSavingsMsg'
Learn More

3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester, Thermo Scientific™

Product Code. 15497106
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 943994-02-3
Molecular Formula C14H18BNO4
Molecular Weight (g/mol) 275.111
MDL Number MFCD12755796
InChI Key ZXOSNHPLTJAXSA-UHFFFAOYSA-N
Synonym 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-benzo b 1,4 oxazin-3 4h-one, 3-oxo-2h,4h-benzo b 1,4 oxazine-6-boronic acid pinacol ester, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-1,4-benzoxazin-3 4h-one, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,4-dihydro-1,4-benzoxazin-3-one, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h, 6-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-1,4-benzoxazin-3-ol, 3-oxo-3,4-dihydro-2h-benzo b 1,4 oxazin-6-yl boronic acid pinacol ester, 6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-2h-benzo b 1,4 oxazin-3 4h-one, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-1,4-benzoxazin-3-ol, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-4h-1,4-benzoxazin-3-one
PubChem CID 46856474
IUPAC Name 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-4H-1,4-benzoxazin-3-one
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)OCC(=O)N3

Specifications

Quantity 1 g
Formula Weight 275.11
Physical Form Solid
Chemical Name or Material 3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'