missing translation for 'onlineSavingsMsg'
Learn More

3-Oxo-3-[4-(trifluoromethoxy)phenyl]propanenitrile, 97%, Thermo Scientific™

Product Code. 10099431
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 122454-46-0
Molecular Formula C10H6F3NO2
Molecular Weight (g/mol) 229.16
MDL Number MFCD00052146
InChI Key IYZQGBBCANKWMX-UHFFFAOYSA-N
Synonym 3-oxo-3-4-trifluoromethoxy phenyl propanenitrile, 4-trifluoromethoxy benzoyl acetonitrile, 4-trifluoromethoxy benzoylacetonitrile, 4-trifluoromethoxybenzoyl acetonitrile, 3-oxo-3-4-trifluoromethoxy phenyl propionitrile, benzenepropanenitrile, beta-oxo-4-trifluoromethoxy, maybridge1_004897, acmc-1bxb1
PubChem CID 2777239
IUPAC Name 3-oxo-3-[4-(trifluoromethoxy)phenyl]propanenitrile
SMILES FC(F)(F)OC1=CC=C(C=C1)C(=O)CC#N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CF3OC6H4C(O)CH2CN
Quantity 1 g
Formula Weight 229.16
Percent Purity 97%
Chemical Name or Material 3-Oxo-3-[4-(trifluoromethoxy)phenyl]propanenitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'