missing translation for 'onlineSavingsMsg'
Learn More

3-(Pentafluorothio)benzoyl chloride, 97%, Thermo Scientific™

Product Code. 15495997
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 401892-81-7
Molecular Formula C7H4ClF5OS
Molecular Weight (g/mol) 266.61
MDL Number MFCD03788522
InChI Key IDHIADOQLRPGHK-UHFFFAOYSA-N
Synonym 3-pentafluorosulfanyl benzoyl chloride, 3-pentafluoro-??-sulfanyl benzoyl chloride, 3-pentafluorothio benzoyl chloride, 3-pentafluorosulfur benzoyl chloride, 3-pentafluoro-$l^ 6-sulfanyl benzoyl chloride, 3-pentafluoro-lambda6-sulfanyl benzoyl chloride
PubChem CID 2779213
IUPAC Name 3-(pentafluoro-$l^{6}-sulfanyl)benzoyl chloride
SMILES FS(F)(F)(F)(F)C1=CC=CC(=C1)C(Cl)=O

Specifications

Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 266.61
Percent Purity 97%
Chemical Name or Material 3-(Pentafluorothio)benzoyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'