missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Pentanone, 98%, pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | -40.0°C |
Color | Colorless |
Density | 0.8150g/mL |
Boiling Point | 100.0°C to 102.0°C |
Flash Point | 7°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass bottle |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0465446 | Certificate of Analysis | 15/08/2024 | 15581044,155810025,155815000 |
A0462902 | Certificate of Analysis | 20/05/2024 | 15581044,155810025,155815000 |
A0395772 | Certificate of Analysis | 14/12/2022 | 15581044,155810025,155815000 |
A0405915 | Certificate of Analysis | 13/12/2022 | 15581044,155810025,155815000 |
A0416649 | Certificate of Analysis | 12/12/2022 | 15581044,155810025,155815000 |
Safety and Handling


Product Identifier
- 3-Pentanone
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H336-May cause drowsiness or dizziness.
- H335-May cause respiratory irritation.
- H319-Causes serious eye irritation.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3-Pentanone, 98%, pure, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction