missing translation for 'onlineSavingsMsg'
Learn More
3-Pentanone, 99%, Thermo Scientific Chemicals Product Code.: 11428166

3-Pentanone, 99%, Thermo Scientific Chemicals

Product Code. 11428166
500 mL
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
This item is not returnable. View return policy

Product Code. 11428166

Brand: Thermo Scientific Alfa Aesar A15297.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3-Pentanone is mainly used as a solvent in paint and a precursor to vitamin E. It is used as a reagent to synthesize ethyl 2-cyano-3,3-diethylacrylate by Knoevenagel condensation. It also shows anticonvulsant effect in several types of mouse seizure models. It serves as an intermediate in the manufacture of pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Pentanone is mainly used as a solvent in paint and a precursor to vitamin E. It is used as a reagent to synthesize ethyl 2-cyano-3,3-diethylacrylate by Knoevenagel condensation. It also shows anticonvulsant effect in several types of mouse seizure models. It serves as an intermediate in the manufacture of pharmaceuticals.

Solubility
Soluble in water, alcohol, ether and carbonterachloride. Slightly miscible with ethanol and acetone.

Notes
Incompatible with strong oxidizing agents, reducing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-22-0
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00009320
InChI Key FDPIMTJIUBPUKL-UHFFFAOYSA-N
Synonym 3-pentanone, diethyl ketone, propione, ethyl ketone, dimethylacetone, metacetone, methacetone, diethylcetone, ethyl propionyl, diethylketoneShow MoreShow Less
PubChem CID 7288
ChEBI CHEBI:67886
IUPAC Name pentan-3-one
SMILES CCC(=O)CC

Specifications

Melting Point -39°C
Density 0.812
Boiling Point 102°C to 103°C
Flash Point 7°C (45°F)
Odor Acetone-like
Refractive Index 1.392
Quantity 500 mL
UN Number UN1156
Beilstein 635749
Merck Index 14,3121
Solubility Information Soluble in water,alcohol,ether and carbonterachloride. Slightly miscible with ethanol and acetone.
Formula Weight 86.13
Percent Purity 99%
Chemical Name or Material 3-Pentanone
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250757 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA15297.AP,A15297.AE,A15297.0F
Lot Number10247157 Certificate TypeCertificate of Analysis Date29/06/2023 Catalog NumberA15297.AP,A15297.AE,A15297.0F,A15297
Lot Number10238834 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA15297.AP,A15297.AE,A15297.0F
Lot Number10183183 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15297.AP,A15297.AE,A15297.0F
Lot NumberH7063B Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15297.AP,A15297.AE,A15297.0F
compliance-icons compliance-icons
Product Identifier
  • 3-Pentanone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contains : Diethyl ketone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Pentanone, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.