missing translation for 'onlineSavingsMsg'
Learn More

3-Pentanone, ReagentPlus™, ≥99.0% (GC), Honeywell™

Product Code. 15641190
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
20 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade,≥99.0% (GC)

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-22-0
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00009320
InChI Key FDPIMTJIUBPUKL-UHFFFAOYSA-N
Synonym 3-pentanone, diethyl ketone, propione, ethyl ketone, dimethylacetone, metacetone, methacetone, diethylcetone, ethyl propionyl, diethylketone
PubChem CID 7288
ChEBI CHEBI:67886
IUPAC Name pentan-3-one
SMILES CCC(=O)CC

Specifications

Melting Point -39°C
Color Colorless
Density 0.813g/mL at 25°C
Boiling Point 102°C
Flash Point 7°C (44.6°F)
Packaging Glass Bottle
Linear Formula CH3CH2COCH2CH3
Refractive Index n20/D 1.392(lit.)
Quantity 1 L
UN Number UN1156
Beilstein 635749
Solubility Information Slightly soluble in water
Formula Weight 86.13g/mol
Percent Purity ≥99.0% (GC)
Grade ReagentPlus™
Chemical Name or Material 3-Pentanone
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'