missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Pentanone, ReagentPlus™, ≥99.0% (GC), Honeywell™
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
20 L
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Melting Point | -39°C |
Color | Colorless |
Density | 0.813g/mL at 25°C |
Boiling Point | 102°C |
Flash Point | 7°C (44.6°F) |
Packaging | Glass Bottle |
Linear Formula | CH3CH2COCH2CH3 |
Refractive Index | n20/D 1.392(lit.) |
Quantity | 2.5 L |
UN Number | UN1156 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'