missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxybenzoyl chloride, ≥97%, Thermo Scientific™

Product Code. 10698283
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 3586-15-0
Molecular Formula C13H9ClO2
Molecular Weight (g/mol) 232.663
MDL Number MFCD03424712
InChI Key TTZXIWBOKOZOPL-UHFFFAOYSA-N
Synonym benzoyl chloride, 3-phenoxy, m-phenoxybenzoyl chloride, m-phenoxy benzoylchoride, 3-phenoxy benzoyl chloride, 3-phenoxy-benzoyl chloride, benzoyl chloride,3-phenoxy
PubChem CID 2760341
IUPAC Name 3-phenoxybenzoyl chloride
SMILES C1=CC=C(C=C1)OC2=CC=CC(=C2)C(=O)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 232.67
Percent Purity 97%
Chemical Name or Material 3-Phenoxybenzoyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'