missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyl-5-piperazino-1,2,4-thiadiazole, 97%, Thermo Scientific™

Product Code. 10586124
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 306935-14-8
Molecular Formula C12H14N4S
Molecular Weight (g/mol) 246.332
MDL Number MFCD00115115
InChI Key UMFMHSLRNJBGKO-UHFFFAOYSA-N
PubChem CID 2777760
IUPAC Name 3-phenyl-5-piperazin-1-yl-1,2,4-thiadiazole
SMILES C1CN(CCN1)C2=NC(=NS2)C3=CC=CC=C3

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Synonym 3-phenyl-5-piperazino-1,2,4-thiadiazole, 1-3-phenyl-1,2,4-thiadiazol-5-yl piperazine, 3-phenyl-5-piperazin-1-yl-1,2,4-thiadiazole, piperazine, 1-3-phenyl-1,2,4-thiadiazol-5-yl, maybridge1_003948, 3-phenyl-5-piperazinyl-1,2,4-thiadiazole, 1-3-phenyl-1,2,4-thiadiazol-5-yl piperazin, 1-3-phenyl-1,2,4-thiadiazol-5-yl-piperazine, 1-3-phenyl-1,2,4 thiadiazol-5-yl-piperazine, piperazine,1-3-phenyl-1,2,4-thiadiazol-5-yl
Formula Weight 246.34
Percent Purity 97%
Chemical Name or Material 3-Phenyl-5-piperazino-1, 2, 4-thiadiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'