missing translation for 'onlineSavingsMsg'
Learn More

3-(Phenylaminomethyl)benzeneboronic acid pinacol ester, 97%, Thermo Scientific™

Product Code. 11337649
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073353-90-8
Molecular Formula C19H24BNO2
Molecular Weight (g/mol) 309.216
MDL Number MFCD09266198
InChI Key FMCOCBRSLHQLQR-UHFFFAOYSA-N
Synonym 3-phenylaminomethyl benzeneboronic acid pinacol ester, n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl aniline, n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl aniline, amtb061, 3-phenylaminomethyl phenylboronic acid pinacol ester, 3-phenylaminomethyl phenyl boronic acid pinacol ester, 3-phenylamino methyl phenylboronic acid, pinacol ester, 2-3-anilinomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl benzenamine
PubChem CID 46739047
IUPAC Name N-[[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]aniline
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)CNC3=CC=CC=C3

Specifications

Melting Point 112°C to 115°C
Quantity 1 g
Formula Weight 309.21
Percent Purity 97%
Chemical Name or Material 3-(Phenylaminomethyl)benzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'