Learn More
3-Phenylazo-2,6-diaminopyridine hydrochloride, 98+%, Thermo Scientific Chemicals
£10.90 - £166.00
Chemical Identifiers
CAS | 136-40-3 |
---|---|
Molecular Formula | C11H12ClN5 |
Molecular Weight (g/mol) | 249.702 |
MDL Number | MFCD00035347 |
InChI Key | QQBPIHBUCMDKFG-UHFFFAOYSA-N |
Synonym | phenazopyridine hydrochloride, phenazopyridine hcl, urodine, pyridium, pyripyridium, bisteril, diridone, mallophene, phenazodine, pyrazodine |
PubChem CID | 8691 |
ChEBI | CHEBI:71419 |
IUPAC Name | 3-phenyldiazenylpyridine-2,6-diamine;hydrochloride |
SMILES | C1=CC=C(C=C1)N=NC2=C(N=C(C=C2)N)N.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11484947
|
Thermo Scientific Alfa Aesar
B21886.03 |
1 g | |||||||
11494947
|
Thermo Scientific Alfa Aesar
B21886.06 |
5 g |
£45.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11404957
|
Thermo Scientific Alfa Aesar
B21886.14 |
25 g |
£166.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
136-40-3 | |
249.702 | |
QQBPIHBUCMDKFG-UHFFFAOYSA-N | |
8691 | |
3-phenyldiazenylpyridine-2,6-diamine;hydrochloride |
C11H12ClN5 | |
MFCD00035347 | |
phenazopyridine hydrochloride, phenazopyridine hcl, urodine, pyridium, pyripyridium, bisteril, diridone, mallophene, phenazodine, pyrazodine | |
CHEBI:71419 | |
C1=CC=C(C=C1)N=NC2=C(N=C(C=C2)N)N.Cl |
Specifications
136-40-3 | |
C11H12ClN5 | |
phenazopyridine hydrochloride, phenazopyridine hcl, urodine, pyridium, pyripyridium, bisteril, diridone, mallophene, phenazodine, pyrazodine | |
C1=CC=C(C=C1)N=NC2=C(N=C(C=C2)N)N.Cl | |
249.702 | |
CHEBI:71419 | |
≥98% |
∼240°C (decomposition) | |
MFCD00035347 | |
QQBPIHBUCMDKFG-UHFFFAOYSA-N | |
3-phenyldiazenylpyridine-2,6-diamine;hydrochloride | |
8691 | |
249.7 | |
3-Phenylazo-2,6-diaminopyridine hydrochloride |
Safety and Handling
GHS H Statement
H301-H334-H351-H317
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing cancer.
May cause an allergic skin reaction.
P201-P202-P261-P264b-P270-P271-P272-P280-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P362-P501c
H302-H315-H317-H319-H335-H350
missing translation for 'einecsNumber' : 205-243-8
missing translation for 'rtecsNumber' : US7875000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only