missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylcinnoline-4-carboxylic acid, 97%, Thermo Scientific™

Catalog Number 10282932
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
10604-21-4
C15H10N2O2
250.257
MFCD00219795
UGJHDXUWDLKCDG-UHFFFAOYSA-N
4-cinnolinecarboxylicacid, 3-phenyl, maybridge1_004872, 3-phenyl-cinnoline-4-carboxylic acid
5709072
3-phenylcinnoline-4-carboxylic acid
C1=CC=C(C=C1)C2=C(C3=CC=CC=C3N=N2)C(=O)O
This item is not returnable. View return policy
10604-21-4
C15H10N2O2
250.257
MFCD00219795
UGJHDXUWDLKCDG-UHFFFAOYSA-N
4-cinnolinecarboxylicacid, 3-phenyl, maybridge1_004872, 3-phenyl-cinnoline-4-carboxylic acid
5709072
3-phenylcinnoline-4-carboxylic acid
C1=CC=C(C=C1)C2=C(C3=CC=CC=C3N=N2)C(=O)O
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 250.26
Percent Purity 97%
Chemical Name or Material 3-Phenylcinnoline-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'