missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3-Phenylisoxazol-5-yl)methylamine, 97%, Thermo Scientific™
£74.80 - £431.00
Chemical Identifiers
CAS | 54408-35-4 |
---|---|
Molecular Formula | C10H10N2O |
Molecular Weight (g/mol) | 174.203 |
InChI Key | AQZLTCXQTOKUAA-UHFFFAOYSA-N |
Synonym | 3-phenylisoxazol-5-yl methanamine, 3-phenyl-5-isoxazolyl methanamine, 3-phenyl-1,2-oxazol-5-yl methanamine, 3-phenylisoxazol-5-yl methylamine, 1-3-phenyl-1,2-oxazol-5-yl methanamine, 5-aminomethyl-3-phenylisoxazole, 1-3-phenylisoxazol-5-yl methanamine, 3-phenyl-5-isoxazolyl methanamine hydrochloride, phenylisoxazolylmethanamine, 3-phenylisoxazol-5-ylmethylamine |
PubChem CID | 2764165 |
IUPAC Name | (3-phenyl-1,2-oxazol-5-yl)methanamine |
SMILES | C1=CC=C(C=C1)C2=NOC(=C2)CN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10737685
|
Thermo Scientific Maybridge
CC64513CB |
250 mg | Amber Glass Bottle |
£74.80
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10452074
|
Thermo Scientific Maybridge
CC64513DA |
1 g | Amber Glass Bottle |
£140.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10794277
|
Thermo Scientific Maybridge
CC64513DE |
5 g | Amber Glass Bottle |
£431.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
54408-35-4 | |
174.203 | |
3-phenylisoxazol-5-yl methanamine, 3-phenyl-5-isoxazolyl methanamine, 3-phenyl-1,2-oxazol-5-yl methanamine, 3-phenylisoxazol-5-yl methylamine, 1-3-phenyl-1,2-oxazol-5-yl methanamine, 5-aminomethyl-3-phenylisoxazole, 1-3-phenylisoxazol-5-yl methanamine, 3-phenyl-5-isoxazolyl methanamine hydrochloride, phenylisoxazolylmethanamine, 3-phenylisoxazol-5-ylmethylamine | |
(3-phenyl-1,2-oxazol-5-yl)methanamine |
C10H10N2O | |
AQZLTCXQTOKUAA-UHFFFAOYSA-N | |
2764165 | |
C1=CC=C(C=C1)C2=NOC(=C2)CN |
Specifications
54408-35-4 | |
C10H10N2O | |
AQZLTCXQTOKUAA-UHFFFAOYSA-N | |
(3-phenyl-1,2-oxazol-5-yl)methanamine | |
2764165 | |
95% |
95% | |
3-phenylisoxazol-5-yl methanamine, 3-phenyl-5-isoxazolyl methanamine, 3-phenyl-1,2-oxazol-5-yl methanamine, 3-phenylisoxazol-5-yl methylamine, 1-3-phenyl-1,2-oxazol-5-yl methanamine, 5-aminomethyl-3-phenylisoxazole, 1-3-phenylisoxazol-5-yl methanamine, 3-phenyl-5-isoxazolyl methanamine hydrochloride, phenylisoxazolylmethanamine, 3-phenylisoxazol-5-ylmethylamine | |
C1=CC=C(C=C1)C2=NOC(=C2)CN | |
174.203 | |
174.2 | |
(3-phenylisoxazol-5-yl)methylamine |
Safety and Handling
GHS Signal Word: Danger