missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylphenol, 90%, Thermo Scientific Chemicals

Product Code. 10567562
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 580-51-8
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.21
MDL Number MFCD00002294
InChI Key UBXYXCRCOKCZIT-UHFFFAOYSA-N
Synonym 3-hydroxybiphenyl, 1,1'-biphenyl-3-ol, biphenyl-3-ol, m-phenylphenol, m-hydroxydiphenyl, 3-biphenylol, m-hydroxybiphenyl, 3-hydroxydiphenyl, 3-phenyl phenol, unii-zu11x47h4o
PubChem CID 11381
ChEBI CHEBI:34338
IUPAC Name 3-phenylphenol
SMILES OC1=CC=CC(=C1)C1=CC=CC=C1

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 72.0°C to 80.0°C
Color Beige-Yellow to White
Boiling Point 300.0°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: slightly soluble. Other solubilities: organic solvents: ethanol,diethyl ether,chlorof,benzene,pyridine
Formula Weight 170.21
Percent Purity 90%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 3-Phenylphenol, 90%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'