Learn More
3-Phenylpropionaldehyde, 95%, Thermo Scientific Chemicals
£40.70 - £465.32
Chemical Identifiers
CAS | 104-53-0 |
---|---|
Molecular Formula | C9H10O |
Molecular Weight (g/mol) | 134.178 |
MDL Number | MFCD00007021 |
InChI Key | YGCZTXZTJXYWCO-UHFFFAOYSA-N |
Synonym | benzenepropanal, 3-phenylpropionaldehyde, hydrocinnamaldehyde, hydrocinnamic aldehyde, dihydrocinnamaldehyde, benzylacetaldehyde, 3-phenyl-1-propanal, 3-phenylpropylaldehyde, 3-phenylpropyl aldehyde, propanal, phenyl |
PubChem CID | 7707 |
IUPAC Name | 3-phenylpropanal |
SMILES | C1=CC=C(C=C1)CCC=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11474173
|
Thermo Scientific Alfa Aesar
A10367.18 |
50 g |
£40.70
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11484173
|
Thermo Scientific Alfa Aesar
A10367.30 |
250 g |
£160.80
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11464173
|
Thermo Scientific Alfa Aesar
A10367.0B |
1000 g |
£465.32
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-Phenylpropionaldehyde is used as a flavoring agent. It is also used in the preparation of cinnamaldehyde, 2-chloro hydrocinnamaldehyde by alfa chlorination and mixture of homopropargyl alcohols. It serves as a precursor for the synthesis of mixture of syn- and anti-beta-hydroxyallylsilanes by hydroboration of allenylsilane. It is also involved in the catalytic asymmetric cyanosilylation to prepare (2S)-2-hydroxy-4-phenylbutanenitrile. Further, it undergoes Henry reaction to form nitroaldols by reacting with nitromethane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Phenylpropionaldehyde is used as a flavoring agent. It is also used in the preparation of cinnamaldehyde, 2-chloro hydrocinnamaldehyde by alfa chlorination and mixture of homopropargyl alcohols. It serves as a precursor for the synthesis of mixture of syn- and anti-beta-hydroxyallylsilanes by hydroboration of allenylsilane. It is also involved in the catalytic asymmetric cyanosilylation to prepare (2S)-2-hydroxy-4-phenylbutanenitrile. Further, it undergoes Henry reaction to form nitroaldols by reacting with nitromethane.
Solubility
Miscible with chloroform, dichloromethane, ethyl acetate, alcohol and ether. Immiscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Chemical Identifiers
104-53-0 | |
134.178 | |
YGCZTXZTJXYWCO-UHFFFAOYSA-N | |
7707 | |
C1=CC=C(C=C1)CCC=O |
C9H10O | |
MFCD00007021 | |
benzenepropanal, 3-phenylpropionaldehyde, hydrocinnamaldehyde, hydrocinnamic aldehyde, dihydrocinnamaldehyde, benzylacetaldehyde, 3-phenyl-1-propanal, 3-phenylpropylaldehyde, 3-phenylpropyl aldehyde, propanal, phenyl | |
3-phenylpropanal |
Specifications
104-53-0 | |
1.017 | |
95°C (203°F) | |
C6H5(CH2)2CHO | |
MFCD00007021 | |
Air Sensitive | |
Miscible with chloroform,dichloromethane,ethyl acetate,alcohol and ether. Immiscible with water. | |
C1=CC=C(C=C1)CCC=O | |
134.178 | |
134.18 | |
3-Phenylpropionaldehyde |
°C to 42°C | |
222°C to 223°C | |
C9H10O | |
1.523 | |
1071910 | |
benzenepropanal, 3-phenylpropionaldehyde, hydrocinnamaldehyde, hydrocinnamic aldehyde, dihydrocinnamaldehyde, benzylacetaldehyde, 3-phenyl-1-propanal, 3-phenylpropylaldehyde, 3-phenylpropyl aldehyde, propanal, phenyl | |
YGCZTXZTJXYWCO-UHFFFAOYSA-N | |
3-phenylpropanal | |
7707 | |
95% |
Safety and Handling
GHS H Statement
H315-H319-H335
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 203-211-8
missing translation for 'rtecsNumber' : MW4890000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
Hazard Statement
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only