missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Phenylpropylamine, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
3-Phenylpropylamine is is a compound used for the synthesis of various pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Phenylpropylamine is is a compound used for the synthesis of various pharmaceuticals.
Solubility
Soluble in Chloroform, Ethyl Acetate. Not miscible in water.
Notes
Store away from air and under dry inert gas. Air sensitive. Incompatible with acids, Acid chlorides, Acid anhydrides, Strong oxidizing agents, Carbon dioxide.
3-Phenylpropylamine is is a compound used for the synthesis of various pharmaceuticals.
Solubility
Soluble in Chloroform, Ethyl Acetate. Not miscible in water.
Notes
Store away from air and under dry inert gas. Air sensitive. Incompatible with acids, Acid chlorides, Acid anhydrides, Strong oxidizing agents, Carbon dioxide.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 0.949 |
Boiling Point | 220°C to 221°C |
Flash Point | 90°C (194°F) |
Odor | Amine-like |
Refractive Index | 1.525 |
Quantity | 10 g |
UN Number | UN2735 |
Beilstein | 2205526 |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in Chloroform,Ethyl Acetate. Not miscible in water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'