missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Phenylpyridine, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
3-Phenylpyridine is a useful synthetic intermediate. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Phenylpyridine is a useful synthetic intermediate. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in chloroform, dichloromethane and ethyl acetate.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
3-Phenylpyridine is a useful synthetic intermediate. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in chloroform, dichloromethane and ethyl acetate.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 1.085 |
Boiling Point | 272°C |
Flash Point | >110°C (230°F) |
Refractive Index | 1.616 |
Quantity | 25 g |
Beilstein | 110400 |
Solubility Information | Soluble in chloroform,dichloromethane and ethyl acetate. |
Formula Weight | 155.2 |
Percent Purity | 97% |
Chemical Name or Material | 3-Phenylpyridine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'