missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 3-Picoline N-oxide, 98%
Click to view available options
Quantity:
500 g
Specifications
Melting Point | 36°C to 40°C |
Density | 1.13 |
Boiling Point | 150°C (15 mmHg) |
Flash Point | 155°C (311°F) |
Quantity | 500 g |
Beilstein | 106327 |
Sensitivity | Hygroscopic |
Formula Weight | 109.13 |
Percent Purity | 98% |
Chemical Name or Material | 3-Picoline N-oxide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'