missing translation for 'onlineSavingsMsg'
Learn More

3-Piperidin-1-ylmethyl benzylamine, ≥97%, Thermo Scientific™

Product Code. 10513423
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 91271-80-6
Molecular Formula C13H20N2
Molecular Weight (g/mol) 204.317
MDL Number MFCD06408781
InChI Key BMNFPARGJHNMAX-UHFFFAOYSA-N
Synonym 3-piperidin-1-ylmethyl-benzylamine, 1-3-piperidin-1-ylmethyl phenyl methanamine, 3-piperidin-1-ylmethyl phenyl methanamine, 3-piperidin-1-ylmethyl benzyl amine, 3-piperidylmethyl phenyl methylamine, 3-piperidin-1-ylmethylbenzylamine, benzenemethanamine, 3-1-piperidinylmethyl, 3-piperidin-1-yl methyl phenyl methanamine, 1-3-piperidin-1-ylmethyl phenyl methylamine
PubChem CID 2794678
IUPAC Name [3-(piperidin-1-ylmethyl)phenyl]methanamine
SMILES C1CCN(CC1)CC2=CC(=CC=C2)CN

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 204.32
Percent Purity 97%
Chemical Name or Material 3-Piperidin-1-ylmethyl benzylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'