Learn More
Fisherbrand™ 3-Ply Sterile Facemasks
3-Ply Sterile Facemasks constructed of polyethylene apertured film and PTFE (polytetrafluoroethylene).
£133.00 - £145.00
Specifications
Product Type | Cleanroom Face Mask |
---|---|
Color | White |
Sterility | Sterile |
Style | Pleated with Ties |
Certifications/Compliance | The masks are rigorously tested and manufactured in an ISO 13485 certified environment. The tests are validated by an independent control laboratory. This product is not a medical device in context of Regulation (EU) 2017/745/Directive 93/42/EEC nor personal protective equipment in context of Regulation (EU) 2016/425. Each batch is sterilized following ISO 11137 and substantiate a sterility assurance level SAL 10−6 according to ISO 11137-2:2013. |
Description
- Constructed from a combination of pleated clear, polyethylene apertured film and PTFE (polytetrafluoroethylene), with soft tie strings
- Manufactured in systems complying ISO 13485:2016 requirements
- Certified Sterile
Specifications
Cleanroom Face Mask | |
Sterile | |
The masks are rigorously tested and manufactured in an ISO 13485 certified environment. The tests are validated by an independent control laboratory. This product is not a medical device in context of Regulation (EU) 2017/745/Directive 93/42/EEC nor personal protective equipment in context of Regulation (EU) 2016/425. Each batch is sterilized following ISO 11137 and substantiate a sterility assurance level SAL 10−6 according to ISO 11137-2:2013. | |
Industrial Use Only |
White | |
Pleated with Ties | |
Single-use |