missing translation for 'onlineSavingsMsg'
Learn More

3-Pyridineacetamide, 99%, Thermo Scientific Chemicals

Product Code. 11461100
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3724-16-1
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.154
MDL Number MFCD08236789
InChI Key YDHIMEXEGOCNHU-UHFFFAOYSA-N
Synonym 2-pyridin-3-yl acetamide, 3-pyridineacetamide, pyridine-3-acetamide, 2-3-pyridyl acetamide, 2-pyridine-3-yl-acetamide, 3-pyridine acetamide, 3-pyridylacetamide, acmc-1cs2v, 2-3-pyridinyl acetamide, 2-pyridin-3-yl acetamid
PubChem CID 564767
IUPAC Name 2-pyridin-3-ylacetamide
SMILES C1=CC(=CN=C1)CC(=O)N

Specifications

Melting Point 122°C to 124°C
Density 1.17
Quantity 5 g
Formula Weight 136.15
Percent Purity 99%
Chemical Name or Material 3-Pyridineacetamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'