missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrrolidin-1-ylbenzoic acid, 97%, Thermo Scientific™

Product Code. 10621465
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 72548-79-9
Molecular Formula C11H13NO2
Molecular Weight (g/mol) 191.23
MDL Number MFCD03426724
InChI Key HRVWGEKZONYEMK-UHFFFAOYSA-N
PubChem CID 651721
IUPAC Name 3-pyrrolidin-1-ylbenzoic acid
SMILES C1CCN(C1)C2=CC=CC(=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 3-pyrrolidin-1-yl benzoic acid, 3-pyrrolidin-1-yl-benzoic acid, 3-1-pyrrolidinyl benzoic acid, benzoic acid, 3-1-pyrrolidinyl, 3-pyrrolidinylbenzoic acid, pubchem19568, 3-pyrrolidinobenzoic acid, 3-1-pyrrolidinyl benzoicacid, 1-3-carboxyphenyl pyrrolidine, benzoic acid,3-1-pyrrolidinyl
Formula Weight 191.23
Percent Purity 97%
Chemical Name or Material 3-Pyrrolidin-1-ylbenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'