Learn More
3-Quinolineboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
£60.70 - £195.00
Chemical Identifiers
CAS | 171364-85-5 |
---|---|
Molecular Formula | C15H18BNO2 |
Molecular Weight (g/mol) | 255.12 |
MDL Number | MFCD05155218 |
InChI Key | ARRJAONCYUAPJR-UHFFFAOYSA-N |
Synonym | 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, quinoline-3-boronic acid pinacol ester, 3-quinolineboronic acid pinacol ester, quinoline-3-boronic acid pinacolate, 3-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, quinoline-3-boronic acid, pinacol ester, quinoline, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, pubchem1882, quinoline-3-boronic acidpinacolate, quinoline-3-boronicacidpinacolester |
PubChem CID | 10848590 |
IUPAC Name | 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline |
SMILES | CC1(C)OB(OC1(C)C)C1=CC2=CC=CC=C2N=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11976501
|
Thermo Scientific Acros
432110010 |
1 g |
£60.70
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11986501
|
Thermo Scientific Acros
432110050 |
5 g |
£195.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
171364-85-5 | |
255.12 | |
ARRJAONCYUAPJR-UHFFFAOYSA-N | |
10848590 | |
CC1(C)OB(OC1(C)C)C1=CC2=CC=CC=C2N=C1 |
C15H18BNO2 | |
MFCD05155218 | |
3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, quinoline-3-boronic acid pinacol ester, 3-quinolineboronic acid pinacol ester, quinoline-3-boronic acid pinacolate, 3-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, quinoline-3-boronic acid, pinacol ester, quinoline, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, pubchem1882, quinoline-3-boronic acidpinacolate, quinoline-3-boronicacidpinacolester | |
3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline |
Specifications
171364-85-5 | |
100.0 | |
Brown to White | |
96% min. (GC) | |
C15H18BNO2 | |
3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, quinoline-3-boronic acid pinacol ester, 3-quinolineboronic acid pinacol ester, quinoline-3-boronic acid pinacolate, 3-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, quinoline-3-boronic acid, pinacol ester, quinoline, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, pubchem1882, quinoline-3-boronic acidpinacolate, quinoline-3-boronicacidpinacolester | |
CC1(C)OB(OC1(C)C)C1=CC2=CC=CC=C2N=C1 | |
255.12 | |
255.13 | |
Powder or Solid |
96.0 | |
55°C to 57°C | |
Authentic | |
Glass Bottle | |
MFCD05155218 | |
ARRJAONCYUAPJR-UHFFFAOYSA-N | |
3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline | |
10848590 | |
97% | |
3-Quinolineboronic acid pinacol ester |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
- 3-Quinolineboronic acid pinacol ester
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only