missing translation for 'onlineSavingsMsg'
Learn More

3-Sulfobenzoic acid disodium salt monohydrate, 97%, Thermo Scientific™

Product Code. 11379829
Click to view available options
Quantity:
25 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14995-40-5
Molecular Formula C7H4O5S
Molecular Weight (g/mol) 200.17
MDL Number MFCD00149379
InChI Key QMWGSOMVXSRXQX-UHFFFAOYSA-L
Synonym sodium 3-sulfonatobenzoate hydrate, 3-sulfobenzoic acid disodium salt monohydrate, disodium 3-sulfonatobenzoate hydrate
PubChem CID 2734925
SMILES [O-]C(=O)C1=CC=CC(=C1)S([O-])(=O)=O

Specifications

Quantity 500 g
IUPAC Name disodium;3-sulfonatobenzoate;hydrate
Formula Weight 264.17 (246.15 Anhydrous)
Percent Purity 97%
Chemical Name or Material 3-Sulfobenzoic acid disodium salt monohydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'