missing translation for 'onlineSavingsMsg'
Learn More

3-Sulfolene, 98%, Thermo Scientific Chemicals

Product Code. 11431588
Click to view available options
Quantity:
100 g
500 g
1000 g
5000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is a source of cisoid butadiene for Diels-Alder reactions. This chemical is also a convenient source of sulfur dioxide. It is a solvent used in the petrochemical industry for the extraction of aromatics from hydrocarbon streams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a source of cisoid butadiene for Diels-Alder reactions. This chemical is also a convenient source of sulfur dioxide. It is a solvent used in the petrochemical industry for the extraction of aromatics from hydrocarbon streams.

Solubility
Soluble in water, ethanol, benzene, ether.

Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-79-2
Molecular Formula C4H6O2S
Molecular Weight (g/mol) 118.15
MDL Number MFCD00005481
InChI Key MBDNRNMVTZADMQ-UHFFFAOYSA-N
Synonym 3-sulfolene, butadiene sulfone, sulfolene, thiophene, 2,5-dihydro-, 1,1-dioxide, beta-sulfolene, 2,5-dihydrothiophene sulfone, 2,5-dihydrothiophene dioxide, 2,5-dihydrothiophene-1,1-dioxide, sulfol-3-ene, ccris 569
PubChem CID 6498
IUPAC Name 2,5-dihydrothiophene 1,1-dioxide
SMILES C1C=CCS1(=O)=O

Specifications

Melting Point 63°C to 67°C
Density 1.314
Flash Point 112°C (233°F)
Quantity 1000 g
Beilstein 107004
Merck Index 14,8956
Solubility Information Soluble in water,ethanol,benzene,ether.
Formula Weight 118.15
Percent Purity 98%
Chemical Name or Material 3-Sulfolene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'