missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(tert-Butyl)-1-methyl-1H-pyrazole-5-carboxylic acid, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 182.22 |
Percent Purity | 95% |
Chemical Name or Material | 3-(tert-Butyl)-1-methyl-1H-pyrazole-5-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'