missing translation for 'onlineSavingsMsg'
Learn More

(3-Thien-3-ylphenyl)methanol, ≥97%, Thermo Scientific™

Product Code. 10341103
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10341103

Brand: Thermo Scientific Maybridge CC48409CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 89929-82-8
Molecular Formula C11H10OS
Molecular Weight (g/mol) 190.26
MDL Number MFCD06203083
InChI Key VSWBZMMHNMDWJL-UHFFFAOYSA-N
Synonym 3-thien-3-ylphenyl methanol, 3-thiophen-3-yl phenyl methanol, 3-3-thienyl phenyl methanol, 3-thiophen-3-ylphenyl methanol, acmc-20ls08, 3-3-thienyl phenylmethanol, 3-3-thiophenyl phenyl methanol, 3-3-thienyl phenyl methan-1-ol
PubChem CID 7164662
IUPAC Name (3-thiophen-3-ylphenyl)methanol
SMILES C1=CC(=CC(=C1)CO)C2=CSC=C2

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 250 mg
Formula Weight 190.27
Percent Purity 95%
Chemical Name or Material (3-Thien-3-ylphenyl)methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.