Learn More
3-Thiophenesulfonyl chloride, ≥95%, Thermo Scientific™
£23.00 - £44.95
Chemical Identifiers
CAS | 51175-71-4 |
---|---|
Molecular Formula | C4H3ClO2S2 |
Molecular Weight (g/mol) | 182.64 |
MDL Number | MFCD02682011 |
InChI Key | YSPWSQNKRBSICH-UHFFFAOYSA-N |
Synonym | 3-thiophenesulfonyl chloride, 3-thiophenesulfonylchloride, chloro-3-thienylsulfone, acmc-209ksa, 3-thienylsulfonyl chloride, 3-thiophenesulphonylchloride, 3-thienyl-sulfonyl chloride, 3-chlorosulphonyl thiophene, 3-thienyl sulfonyl chloride, 3-thiophene sulfonyl chloride |
PubChem CID | 2776378 |
IUPAC Name | thiophene-3-sulfonyl chloride |
SMILES | ClS(=O)(=O)C1=CSC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10394082
|
Thermo Scientific Maybridge
CC13103CB |
250 mg |
£23.00
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10623783
|
Thermo Scientific Maybridge
CC13103DA |
1 g |
£44.95
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
51175-71-4 | |
182.64 | |
YSPWSQNKRBSICH-UHFFFAOYSA-N | |
2776378 | |
ClS(=O)(=O)C1=CSC=C1 |
C4H3ClO2S2 | |
MFCD02682011 | |
3-thiophenesulfonyl chloride, 3-thiophenesulfonylchloride, chloro-3-thienylsulfone, acmc-209ksa, 3-thienylsulfonyl chloride, 3-thiophenesulphonylchloride, 3-thienyl-sulfonyl chloride, 3-chlorosulphonyl thiophene, 3-thienyl sulfonyl chloride, 3-thiophene sulfonyl chloride | |
thiophene-3-sulfonyl chloride |
Specifications
51175-71-4 | |
100.0 | |
C4H3ClO2S2 | |
3-thiophenesulfonyl chloride, 3-thiophenesulfonylchloride, chloro-3-thienylsulfone, acmc-209ksa, 3-thienylsulfonyl chloride, 3-thiophenesulphonylchloride, 3-thienyl-sulfonyl chloride, 3-chlorosulphonyl thiophene, 3-thienyl sulfonyl chloride, 3-thiophene sulfonyl chloride | |
ClS(=O)(=O)C1=CSC=C1 | |
182.64 | |
182.65 | |
3-Thiophenesulfonyl chloride |
96.5 | |
97% | |
MFCD02682011 | |
YSPWSQNKRBSICH-UHFFFAOYSA-N | |
thiophene-3-sulfonyl chloride | |
2776378 | |
97% |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger