missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethoxy)phenol, 98%, Thermo Scientific Chemicals

Product Code. 11489636
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 827-99-6
Molecular Formula C7H5F3O2
Molecular Weight (g/mol) 178.11
MDL Number MFCD00040987
InChI Key UWLJERQTLRORJN-UHFFFAOYSA-N
Synonym 3-trifluoromethoxy phenol, m-trifluoromethoxy phenol, phenol, 3-trifluoromethoxy, 3-trifluoromethoxy-phenol, 3-hydroxyphenyl trifluoromethyl ether, pubchem1505, m-trifluoromethoxyphenol, pubchem10391
PubChem CID 2733261
IUPAC Name 3-(trifluoromethoxy)phenol
SMILES OC1=CC=CC(OC(F)(F)F)=C1

Specifications

Density 1.379
Boiling Point 68°C to 69°C (12 mmHg)
Flash Point 84°C (183°F)
Refractive Index 1.446
Quantity 1 g
Beilstein 1868036
Formula Weight 178.11
Percent Purity 98%
Chemical Name or Material 3-(Trifluoromethoxy)phenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'