missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethoxy)phenol, 98%, Thermo Scientific™

Product Code. 10115950
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 827-99-6
Molecular Formula C7H5F3O2
Molecular Weight (g/mol) 178.11
MDL Number MFCD00040987
InChI Key UWLJERQTLRORJN-UHFFFAOYSA-N
Synonym 3-trifluoromethoxy phenol, m-trifluoromethoxy phenol, phenol, 3-trifluoromethoxy, 3-trifluoromethoxy-phenol, 3-hydroxyphenyl trifluoromethyl ether, pubchem1505, m-trifluoromethoxyphenol, pubchem10391
PubChem CID 2733261
IUPAC Name 3-(trifluoromethoxy)phenol
SMILES OC1=CC=CC(OC(F)(F)F)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.3700g/mL
Boiling Point 69.0°C to 70.0°C (12.0 mmHg)
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CF3OC6H4OH
Refractive Index 1.4450 to 1.4470
Quantity 1 g
Specific Gravity 1.37
Formula Weight 178.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethoxy)phenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'