missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[3-(Trifluoromethyl)phenyl]methanethiol, Tech., Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Specifications
Packaging | Amber Glass Bottle |
Linear Formula | CF3C6H4CH2SH |
Quantity | 1 g |
Formula Weight | 192.2 |
Percent Purity | Tech. |
Grade | Technical |
Chemical Name or Material | [3-(Trifluoromethyl)phenyl]methanethiol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'